Läs mer
Tocris Bioscience™ Janelia Fluor™ 635b, Maleimide
Beskrivning
Spontaneously Blinking Janelia Fluor™ Dyes allow facile single-molecule localization microscopy (SMLM) in cells and dense biomolecular structures, without the need for photoactivation or redox buffers. These spontaneously blinking dyes harness Janelia Fluor™ technology to delivery dyes that automatically cycle between 'off' and 'on' states with an ideal duty cycle for super resolution microscopy experiments.
Key Information: Janelia Fluor™ 635b, Maleimide is a spontaneously blinking fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity) for the labeling of protein.
Used for: suitable to characterize the physical properties of dense biomolecule condensates and complex macromolecular assemblies.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: derived from Janelia Fluor™ 635, Maleimide (Cat. No. 8027). Excitation and emission maxima (λ) are 651 nM and 667 nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-3,7-bis(3-fluoroazetidin-1-yl)-5,5-dimethyl-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxamide |
| Kvantitet | 200 μg |
| Mål | Sulfhydyl-Reactive Janelia Fluor Dyes |
| Molekylformel | C35H34F2N4O4Si |
| Renhet | 0.9 |