Läs mer
Tocris Bioscience™ Janelia Fluor™ 646, Haloalkane
Beskrivning
Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, light sheet microscropy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). Stable to fixation. Cell permeable.
Properties and Photophysical Data: Janelia Fluor™ 646, Haloalkane shows a high degree of fluorogenicity. Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); lactone-zwitterion equilibrium constant (KL-Z) = 0.0012. Exhibits low non-specific background staining.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2-((6-chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)benzoate |
| CAS | 1811539-60-2 |
| Kvantitet | 10 μg |
| Mål | Janelia Fluor Dyes |
| Molekylformel | C39H48ClN3O5Si |
| Fysisk form | Solid |
| Färg | Off-white |
| Produkttyp | Bioactive Small Molecule |
| Renhet | 0.95 |