missing translation for 'onlineSavingsMsg'
Läs mer

Jatrorrhizine Hydroxide, TRC

Produktkod. 30006288
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006288 10 mg
30003451 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30006288 Leverantör Toronto Research Chemicals Leverantörsnummer TRCJ21090010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 483-43-2
Molekylformel C20H21NO5
Molekylvikt (g/mol) 355.38
Synonym 7,8,13,13a-tetradehydro-3-hydroxy-2,9,10-trimethoxy-Berbinium, hydroxide, Neprotine hydroxide
IUPAC-namn 2,9,10-trimethoxy-5,6-dihydroisoquinolino[3,2-a]isoquinolin-7-ium-3-ol;hydroxide
LEDER OC1=C(OC)C=C2C(CC[N+]3=C2C=C(C=CC(OC)=C4OC)C4=C3)=C1.[OH-]

Specifikationer

Kemiskt namn eller material Jatrorrhizine Hydroxide
Kvantitet 10 mg
InChI formel InChI=1S/C20H19NO4.H2O/c1-23-18-5-4-12-8-16-14-10-19(24-2)17(22)9-13(14)6-7-21(16)11-15(12)20(18)25-3;/h4-5,8-11H,6-7H2,1-3H3;1H2
Formel vikt 355.142
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.