Learn More
Cole-Parmer™ Jenway™ Glass Cuvettes
Brand:  Cole-Parmer™ 83070-40
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13131734
  • 2071.54 SEK / Each
Estimated Shipment: 07-10-2024
to see stock.
Specifications
Cuvette
7.0 mL
Glass
20 mm