Learn More
Cole-Parmer™ Jenway™ PH Electrode Storage Solution
Product Code. 11490479
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
150mL
Product Code. Quantity Unit Size
11490479 250 mL 150mL
1 options
Product Code. 11490479 Supplier Cole-Parmer™ Supplier No. 9995960
  • ONLINE EXCLUSIVE PRICE
    621.00 SEK
    556.11 SEK / 150ml
    Save 64.89 SEK
    10% Av
 Restricted Item
Beräknad leverans: 18-12-2026
Optimize the performance of your Jenway meter
  • Easy to install
Specifications
Filling Solution Type pH Electrode Storage
Quantity 250 mL