Läs mer
Jervine, TRC
Produktkod. 30011777
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011777 1 mg
30004729 5 mg
30007390 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCJ2110001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 469-59-0
Molekylformel C27 H39 N O3
Molekylvikt (g/mol) 425.6
Synonym Spiro[9H-benzo[a]fluorene-9,2'(3'H)-furo[3,2-b]pyridin]-11(1H)-one, 2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-hexadecahydro-3-hydroxy-3',6',10,11b-tetramethyl-, (2'R,3S,3'R,3'aS,6'S,6aS,6bS,7'aR,11aS,11bR)-, Jervine (8CI), Veratraman-11-one, 17,23-epoxy-3-hydroxy-, (3β,23β)-, (2'R,3S,3'R,3'aS,6'S,6aS,6bS,7'aR,11aS,11bR)-2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-Hexadecahydro-3-hydroxy-3',6',10,11b-tetramethylspiro[9H-benzo[a]fluorene-9,2'(3'H)-furo[3,2-b]pyridin]-11(1H)-one, Jervin, NSC 23898, NSC 7520, Spiro[9H-benzo[a]fluorene-9,2'(3'H)-furo[3,2-b]pyridin]-11(1H)-one, 2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-hexadecahydro-3-hydroxy-3',6',10,11b-tetramethyl-, [3S-(3α,6aα,6bβ,9α(3'S*,3'aR*,6'R*,7'aS*),11aβ,11bα)]-
IUPAC-namn (3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3-hydroxy-3',6',10,11b-tetramethylspiro[1,2,3,4,6,6a,6b,7,8,11a-decahydrobenzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-11-one
LEDER C[C@@H]1CN[C@@H]2[C@@H](C1)O[C@]3(CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5C(=O)C4=C3C)[C@@H]2C
Specifikationer
Kemiskt namn eller material Jervine
Kvantitet 1 mg
InChI formel InChI=1S/C27H39NO3/c1-14-11-21-24(28-13-14)16(3)27(31-21)10-8-19-20-6-5-17-12-18(29)7-9-26(17,4)23(20)25(30)22(19)15(27)2/h5,14,16,18-21,23-24,28-29H,6-13H2,1-4H3/t14-,16+,18-,19-,20-,21+,23+,24-,26-,27-/m0/s1
Formel vikt 425.293