Läs mer
Tocris Bioscience™ JFX™ 650, NHS ester
Beskrivning
Application: Suitable for confocal microscopy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells) and STED. Cell permeable.
Properties and Photophysical Data: JFX™ 650, NHS ester shows enhanced brightness, photostability and chromostability. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Excitation and emission maxima (λ) are 650 nm and 667 nm, respectively; quantum yield = 0.53; extinction coefficient = 17,600 M-1cm-1; lactone-zwitterion equilibrium constant (KL-Z) = 0.015.
Please see the protocol for further information on protein/antibody labeling and conjugation.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals Biologicals.
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
JFX is a trademark owned by the Howard Hughes Medical Institute.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-(5,5-Dimethyl-3-(pyrrolidin-1-ium-1-ylidene-d8)-7-(pyrrolidin-1-yl-d8)-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-(((2,5-dioxopyrrolidin-1-yl)oxy)carbonyl)benzoate |
| CAS | 2396479-84-6 |
| Kvantitet | 200 μg |
| Mål | Amine-Reactive Janelia Fluor Dyes |
| Molekylformel | C35H19D16N3O6Si |
| Renhet | 0.9 |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.