Läs mer
Tocris Bioscience™ JH 295
Irreversible Nek2 inhibitor
Brand: Tocris Bioscience™ 4322/2
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
JH 295 is an irreversible Nek2 inhibitor (IC50=770 nM). Inhibits Nek2 via alkylation of residue Cys22. Displays 5-fold selectivity for Nek2 over Cdk1. Exhibits no significant effect on Aurora B or Plk1 kinases, bipolar spindle assembly or spindle assembly checkpoint. Cell permeable.Specifikationer
| N-[(3Z)-3-[(2-Ethyl-4-methyl-1H-imidazol-5-yl)methylene]-2,3-dihydro-2-oxo-1H-indol-5-yl]-2-propynamide | |
| 1311143-71-1 | |
| >98% | |
| 2 mg |