Läs mer
Tocris Bioscience™ JH-II-127
Beskrivning
JH-II-127 is a potent and selective LRRK2 inhibitor (IC50 values are 2.2, 6.6 and 47.7 nM for G2019S-mutant, wild-type and A2016T-mutant LRRK2, respectively). Selective for LRRK2 over a panel of 138 kinases. Brain penetrant and orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | [4-[[5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-4-morpholinyl-methanone |
| CAS | 1700693-08-8 |
| Analysprocentintervall | >98% |
| Kvantitet | 50 mg |
| Molekylformel | C19H21ClN6O3 |
| Formel vikt | Observed MW: 416.87 |