Learn More
Cole-Parmer™ Jiffy-Jack Lab Jack
Product Code. 11719128 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (English):
132 lb.
Unit Size:
Each
Product Code. Capacity (English) Unit Size
11719128 132 lb. Each
1 options
Product Code. 11719128 Supplier Cole-Parmer™ Supplier No. 0805720
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Beräknad leverans: 19-01-2027
Jiffy-Jack Lab Jack features excellent side-to-side rigidity and support.
Jiffy-Jack Lab Jack features excellent side-to-side rigidity and support.
  • Provide stable, height-adjustable support for hot plates, baths, glassware, and many other items
  • Positioners are constructed of rugged, chemical-resistant anodized aluminum with stainless steel operating screw.
  • Top-plate skid pad prevents slipping.
  • Each positioner is precision engineered for smooth, continuous operation throughout its range.
  • Easily set the exact height needed with the turn of a knob.
  • A 10 mm tapped hole in the right rear corner accommodates a frame rod (not included) for mounting ring stands and other lab apparatus
  • Maximum support static load is 132 lb. (60 kg)
Specifications
Material Aluminum (Positioner), Stainless Steel (Operating Screw)
Dimensions (L x W) 6 x 6 in. (Top)
Capacity (English) 132 lb.
Quantity 1 Ea.