Läs mer
JNJ-38877605, MedChemExpress
Produktkod. 16493997
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16493997 50 mg -
16450548 5 mg -
16408088 10 mg 10mg
16407856 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16493997 Leverantör Medchem Express Leverantörsnummer HY50683
MedChemExpress JNJ-38877605 is an ATP-competitive inhibitor of c-Met with IC50 of 4 nM, 600-fold selective for c-Met than 200 other tyrosine and serine-threonine kinases.
JNJ-38877605 is an ATP-competitive inhibitor of c-Met with IC50 of 4 nM, 600-fold selective for c-Met than 200 other tyrosine and serine-threonine kinases.
Specifikationer
Kemiskt namn eller material JNJ-38877605
Fysisk form Solid
CAS 943540-75-8
Färg Light Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H13F2N7
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : ≥ 30 mg/mL (79.50 mM)
LEDER FC(F)(C1=NN=C2C=CC(C3=CN(C)N=C3)=NN21)C4=CC5=C(N=CC=C5)C=C4
Molekylvikt (g/mol) 377.35
Formel vikt 377.35
Procent renhet 95.18%