Läs mer
Tocris Bioscience™ JTE 013
:
10mg
Beskrivning
JTE 013 is a sphingosine-1-phosphate (S1P) receptor antagonist, highly selective for S1P2 (EDG-5). Inhibits S1P binding to human S1P2 receptors with an IC50 value of 17.6 nM. At concentrations up to 10 μM, displays 4.2% inhibition of S1P3 and does not antagonize S1P1. Enhances S1P-induced angiogenesis in vivo.
Specifikationer
Specifikationer
| Kvantitet | 10 mg |
| Produkttyp | JTE 013 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 408.29 |
| Inhibitorer | JTE 013 |
For Research Use Only