Learn More
JUN Mouse anti-Human, Mouse, Rat, Clone: 1G6B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66313-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is raised against a region of human JUN. Both phosphorylated (p-c-Jun) and unphosphorylated forms of c-Jun, with sizes of 42-45 kDa and 36-39 kDa, respectively are obtain in some experiments.
Fos and Jun dimerize to form Activator Protein-1 (AP-1), a transcriptional factor that binds to the 12-O-tetradecanoylphorbol 13-acetate (TPA) response element (TRE) of several cellular and viral genes including human collagenase, metallothionein IIa, stromelysin, interleukin 2, SV40 and polyoma. Fos and Jun contain the ′leucine-zipper′ motif that allows for dimerization and an adjacent basic domain required for biological activity. The functionally active form of Fos is in a heterodimer with a member of the Jun family. While Jun family members can form functional homodimers, studies indicate that Fos family members do not self-associate and therefore do not bind DNA on their own. The various dimers differ in their ability to transactivate AP-1 dependent genes.Specifications
JUN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05412, P05627, P17325 | |
Jun | |
JUN Fusion Protein Ag17639 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
1G6B11 | |
Unconjugated | |
JUN | |
Activator protein 1, AP 1, AP1, c Jun, JUN, jun oncogene, P39, Proto oncogene c Jun, Transcription factor AP 1 | |
Mouse | |
Protein G | |
RUO | |
16476, 24516, 3725 | |
-20°C | |
Liquid |