Läs mer
JZL195, MedChemExpress
Produktkod. 16496686
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496686 100 mg -
16456398 10 mg 10mg
16442507 50 mg -
16438987 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496686 Leverantör Medchem Express Leverantörsnummer HY15250
MedChemExpress JZL195 is a selective and efficacious dual fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL) inhibitor with IC50s of 2 and 4 nM, respectively.
JZL195 is a selective and efficacious dual fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL) inhibitor with IC50s of 2 and 4 nM, respectively.
Specifikationer
Kemiskt namn eller material JZL195
CAS 1210004-12-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H23N3O5
Kvantitet 100 mg
Löslighetsinformation DMSO : 50 mg/mL (115.35 mM; Need ultrasonic)
LEDER O=C(OC1=CC=C([N+]([O-])=O)C=C1)N(CC2)CCN2CC3=CC(OC4=CC=CC=C4)=CC=C3
Molekylvikt (g/mol) 433.46
Formel vikt 433.46
Procent renhet 99.81%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation