Läs mer
K134, MedChemExpress
Produktkod. 16452428
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16452428 5 mg -
16430229 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16452428 Leverantör Medchem Express Leverantörsnummer HYU00186
MedChemExpress K134 is a phosphodiesterase 3 (PDE3) inhibitor. The IC50s of K134 toward PDE3A, PDE3B, PDE5, PDE2 and PDE4 are 0.1, 0.28, 12.1, >300 and >300 μM, respectively.
K134 is a phosphodiesterase 3 (PDE3) inhibitor. The IC50s of K134 toward PDE3A, PDE3B, PDE5, PDE2 and PDE4 are 0.1, 0.28, 12.1, >300 and >300 μM, respectively.
Specifikationer
Kemiskt namn eller material K134
CAS 189362-06-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C22H29N3O4
Kvantitet 5 mg
Synonym OPC33509
Löslighetsinformation DMSO : 50 mg/mL (125.16 mM; Need ultrasonic)
LEDER O=C(NCCCOC1=CC2=C(NC(C=C2)=O)C=C1)N(C3CC3)[C@H]4[C@H](O)CCCC4
Molekylvikt (g/mol) 399.48
Formel vikt 399.48
Procent renhet 99.67%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation