Läs mer
Kaempferol-3-O-robinoside-7-O-glucoside, TRC
Produktkod. 30135232
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135232 10 mg
30136799 1 mg
17959854 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK10004010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114924-89-9
Molekylformel C33H40O20
Molekylvikt (g/mol) 756.66
Synonym 3-[[6-O-(6-Deoxy-alpha-L-mannopyranosyl)-beta-D-galactopyranosyl]oxy]-7-(beta-D-glucopyranosyloxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
IUPAC-namn 5-hydroxy-2-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
LEDER C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)cc(O)c4C3=O)c6ccc(O)cc6)[C@H](O)[C@@H](O)[C@H]2O)[C@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Kaempferol-3-O-robinoside-7-O-glucoside
Kvantitet 10 mg
InChI formel InChI=1S/C33H40O20/c1-10-19(37)23(41)26(44)31(48-10)47-9-17-21(39)25(43)28(46)33(52-17)53-30-22(40)18-14(36)6-13(49-32-27(45)24(42)20(38)16(8-34)51-32)7-15(18)50-29(30)11-2-4-12(35)5-3-11/h2-7,10,16-17,19-21,23-28,31-39,41-46H,8-9H2,1H3/t10-,16+,17+,19-,20+,21-,23+,24-,25-,26+,27+,28+,31+,32+,33-/m0/s1
Formel vikt 756.2113