missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Kaiso Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588811
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: PCRYCEKVFP LAEYRTKHEI HHTGERRYQC LACGKSFINY QFMSSHIKSV HSQDPSGDSK LYRLHPCRSL QIRQYAYLSD RSSTIPAMKD DGIGYKVDTG KEPPVGTTTS TQNKPMTWED IFIQQENDSI FKQNVTDGST EFEFIIPESY; Positive Samples: A-431, Mouse kidney; Cellular Location: Cytoplasm, Nucleus.
This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensusKAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
Specifikationer
| Kaiso | |
| Polyclonal | |
| Unconjugated | |
| ZBTB33 | |
| AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10009, 56805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86T24, Q8BN78 | |
| ZBTB33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 523-672 of human Kaiso/Kaiso/ZBTB33 (NP_0067681). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering