Läs mer
Invitrogen™ Kappa Light Chain Monoclonal Antibody (TB28-2), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 17-9970-42
Beskrivning
Description: The monoclonal antibody TB28-2 recognizes the kappa light chain of human immunoglobulin. Approximately 22-kDa, the Ig kappa chain is comprised of one variable and one constant region. Two kappa chains together with the heavy chain comprise the whole Ig structure. Expression of Ig kappa is typically found on more than half of B lymphocytes. Applications Reported: This TB28-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TB28-2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Specifikationer
| Kappa Light Chain | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P01601, P01602, P01834, P0DOX7 | |
| IGK, IGKC, IGKV1-5, IGKV1D-16 | |
| Affinity chromatography | |
| RUO | |
| 28299, 28901, 3514, 50802 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| TB28-2 | |
| APC | |
| IGK | |
| D4Mit8; HCAK1; ig kappa chain c region; Igk; Igk@; Igkc; Igk-C; IGKCD; IGKV; IGKV15; IGKV1-5; Immunoglobulin K; immunoglobulin kappa chain complex; immunoglobulin kappa constant; immunoglobulin kappa light chain; immunoglobulin kappa locus; immunoglobulin kappa variable 1-5; kappa; Kappa LC; Km; L12; L12a; LOC500183; similar to NGF-binding Ig light chain; V1 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.