missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KCNG1 (Human) Recombinant Protein
Human KCNG1 full-length ORF ( AAH06367, 1 a.a. - 275 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH06367 |
---|---|
Gene ID (Entrez) | 3755 |
Name | potassium voltage-gated channel, subfamily G, member 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167091
|
Abnova™
H00003755-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177091
|
Abnova™
H00003755-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MTLLPGDNSDYDYSALSCTSDASFHPAFLPQRQAIKGAFYRRAQRLRPQDEPRQGCQPEDRRRRIIINVGGIKYSLPWTTLDEFPLTRLGQLKACTNFDDILNVCDDYDVTCNEFFFDRNPGAFGTILTFLRAGKLRLLREMCALSFQEELLYWGIAEDHLDGCCKRRYLQKIEEFAEMVEREEEDDALDSEGRDSEGPAEGEGRLGRCMRRLRDMVERPHSGLPGKVFACLSVLFVTVTAVNLSVSTLPSLREEEEQVRAHAPRGNAPPRGKGL
Specifications
AAH06367 | |
potassium voltage-gated channel, subfamily G, member 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
K13, KCNG, KV6.1, MGC12878, kH2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3755 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KCNG1 | |
GST |