missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ KCNG3 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16613145 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16613145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16613145 Leverantör Invitrogen™ Leverantörsnummer PA5106704

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNG3.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen KCNG3
Användningsområden Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen KCNG3
Genaccessionsnr. P59053, Q8R523, Q8TAE7
Gene Alias KCNG3; Kv10.1; Kv10.1a; Kv10.1b; Kv6.3; potassium channel, voltage gated modifier subfamily G, member 3; potassium voltage-gated channel modifier subfamily G member 3; potassium voltage-gated channel subfamily G member 3; potassium voltage-gated channel subfamily G memeber 3; potassium voltage-gated channel, subfamily G, member 3; potassium voltage-gated channel, subfamily G, memeber 3; voltage-gated potassium channel 6.3; voltage-gated potassium channel Kv10.1; voltage-gated potassium channel subunit Kv10.1; Voltage-gated potassium channel subunit Kv6.3; voltage-gated potassium channel subunit Kv6.4
Gensymboler KCNG3
Värd art Rabbit
Immunogen A synthesized peptide derived from human KCNG3(Accession Q8TAE7), corresponding to amino acid residues V181-W231.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 170850, 171011, 225030
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.