missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ KCNIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541041
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: FSSLQTKQRR PSKDKIEDEL EMTMVCHRPE GLEQLEAQTN FTKRELQVLY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.
Specifikationer
| KCNIP1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnip1 | |
| ABP; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; Kchip1.2; KCHIP2; Kcnip1; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; potassium channel-interacting protein 1; potassium voltage-gated channel interacting protein 1; VABP; Vesicle APC-binding protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 30820 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NZI2 | |
| Kcnip1 | |
| Synthetic peptide directed towards the N-terminal of human KCNIP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering