missing translation for 'onlineSavingsMsg'
Learn More

KCNK18 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23712-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882415

  • 1355.00 SEK / 20µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can be used to detect endogenous KCNK18, however, the reason for the additional band of 130-140 kDa is still unknown.

TRESK (TWIK-related spinal cord K+ channel) is a two-pore domain K+ channel that induces outward rectification and functions as a background K+ channel that is abundantly expressed in the spinal cord, thymus, spleen, and leukemic T-lymphocytes. TRESK is activated by increased cytoplasmic calcium concentration through calcineurin as well as by acetylcholine and histamine. It has been suggested that TRESK is linked to acute and chronic pain by activation of calcineurin. Other studies indicate that TRESK may play a role in the regulation of T-cell function and could be an excellent target to treat T-cell originated immune dysfunction.
Specifications

Specifications

KCNK18
Polyclonal
Unconjugated
KCNK18
K2p18.1, KCNK18, TRESK, TRESK 2, TRESK2, TRIK
Rabbit
Antigen Affinity Chromatography
RUO
338567
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z418
KCNK18
KCNK18 Fusion Protein Ag20514
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers