missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KCNN1 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837944
|
Proteintech
17929-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16827944
|
Proteintech
17929-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. KCNN1 is a member of the KCNN family of potassium channels.Specifications
KCNN1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P70606, Q92952, Q9EQR3 | |
3780, 54261, 84036 | |
KCNN1 Fusion Protein Ag12357 | |
Primary | |
-20°C | |
KCNN1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hSK1, KCa2.1, KCNN1, SK, SK1, SKCa 1, SKCA1 | |
KCNN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |