Learn More
KD Scientific Centrifan™ PE Personal Evaporator/Condenser Accessory, Rotors
For use with Centrifan™ PE
Brand:  KD Scientific 788402
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15560540
  • 12825.22 SEK / Each
Estimated Shipment
29-12-2020
Add to basket
Specifications
Centrifan™ PE
Centrifan™ Rotor
6 x 16 x 100mm Test tube and 1 Polypropylene splitter