Learn More
KD Scientific Splitters
Directs airflow into and out of vials to facilitate solvent evaporation
Brand:  KD Scientific 788416
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15580550
  • 435.25 SEK / Each
Estimated Shipment
28-12-2020
Add to basket
Description
Description
  • For use with Centrfian micro centrifuge vial rotor/micro test tube rotor/vial rotor/scintillating vial rotor/10 HPLC vial rotor
Specifications
Specifications
Polyethylene
Centrfian™ 10 HPLC 1.5mL vial rotor
Splitter