Learn More
KD Scientific Splitters 6x20mL
For use with Centrifian 6 × 20mL scintillation vial rotor
POA
Specifications
For Use With (Equipment) Centrfian™ 6 x 20mL scintillation vial
Type Splitter
Material Polyethylene
View More Specs
Description
Description
  • Made of polypropolene
  • Splitter directs airflow into and out of vials to facilitate solvent evaporation
Specifications
Specifications
Centrfian™ 6 x 20mL scintillation vial
Splitter
Polyethylene