Learn More
KD Scientific Splitters 6x20mL
For use with Centrifian 6 × 20mL scintillation vial rotor
POA
Specifications
For Use With (Equipment) | Centrfian™ 6 x 20mL scintillation vial |
---|---|
Type | Splitter |
Material | Polyethylene |
Description
- Made of polypropolene
- Splitter directs airflow into and out of vials to facilitate solvent evaporation
Specifications
Centrfian™ 6 x 20mL scintillation vial | |
Splitter | |
Polyethylene |