Learn More
KD Scientific Splitters 6x20mL
For use with Centrifian 6 × 20mL scintillation vial rotor
Brand:  KD Scientific 788408
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15510550
  • 240.00 SEK / Pack of 5
Estimated Shipment
16-08-2018
Add to basket
Description
Description
  • Made of polypropolene
  • Splitter directs airflow into and out of vials to facilitate solvent evaporation
Specifications
Specifications
Centrfian™ 6 x 20mL scintillation vial
Splitter
Polyethylene
5/Pk.
Videos