Läs mer
Invitrogen™ KDELR1 Monoclonal Antibody (KR-10), APC
GREENER_CHOICE
Produktkod. 17856722
Change view
Kvantitet:
100 μg
Produktkod. Kvantitet
17856722 100 μg
1 options
Produktkod. 17856722 Leverantör Invitrogen™ Leverantörsnummer MA545149
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Mouse Monoclonal Antibody
1 μg/mL was sufficient for detection of KDEL receptor in 20 μg monkey Vero cell lysate by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:AP as the secondary. Detects approximately 25kDa.
KDELR1 (KDEL Endoplasmic Reticulum Protein Retention Receptor 1) is a Protein Coding gene. Diseases associated with KDELR1 include Syndromic X-Linked Intellectual Disability 94 and Myopathy, Centronuclear, 1. Among its related pathways are Transport to the Golgi and subsequent modification and Activation of cAMP-Dependent PKA. Gene Ontology (GO) annotations related to this gene include ER retention sequence binding and KDEL sequence binding. An important paralog of this gene is KDELR2.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen KDELR1
Användningsområden Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona KR-10
Koncentration 1 mg/mL
Konjugera APC
Formulering 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.2
Gen KDELR1
Genaccessionsnr. P24390, P33946, Q569A6, Q99JH8
Gensymboler KDELR1
Värd art Mouse
Immunogen A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH.
Reningsmetod Protein G
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 100349892, 100520962, 100750912, 101122047, 10945, 361577, 484406, 618184, 68137
Målarter Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig, Chicken, Chicken, Frog
Innehåll och lagring 4°C
Produkttyp Antibody
Form Liquid
Isotyp IgG1