missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24311-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.Specifications
KDM2A | |
Polyclonal | |
Unconjugated | |
KDM2A | |
CXXC8, F box protein FBL7, F box protein Lilina, F box/LRR repeat protein 11, FBL11, FBL7, FBXL11, JHDM1A, KDM2A, KIAA1004, LILINA, lysine (K) demethylase 2A, Lysine specific demethylase 2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225876, 22992 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P59997, Q9Y2K7 | |
Kdm2a | |
KDM2A Fusion Protein Ag19448 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |