missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KDM2A |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822795
|
Proteintech
24311-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812795
|
Proteintech
24311-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.Specifications
KDM2A | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P59997, Q9Y2K7 | |
225876, 22992 | |
KDM2A Fusion Protein Ag19448 | |
Primary | |
-20°C | |
KDM2A |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CXXC8, F box protein FBL7, F box protein Lilina, F box/LRR repeat protein 11, FBL11, FBL7, FBXL11, JHDM1A, KDM2A, KIAA1004, LILINA, lysine (K) demethylase 2A, Lysine specific demethylase 2A | |
Kdm2a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |