missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KDR (VEGFR2), His Tag Recombinant Protein
Human KDR (VEGFR2), His Tag Recombinant Protein
5220.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_002244 |
---|---|
Gene Alias | KDR (VEGFR2), VEGFR |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11523497
|
Invitrogen™
PV3660 |
10μg |
5220.00 SEK
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10707763
|
Invitrogen™
PR5992A |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428463
|
Invitrogen™
PR5992C |
100 μg |
18160.00 SEK
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KDR (VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors.Specifications
NP_002244 | |
Human | |
Baculovirus |
KDR (VEGFR2), VEGFR | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.