Learn More
Hamilton™ Custom Kel-F Hub Needles
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
Brand:  Hamilton™ 7750-17
Additional Details : Weight : 1.00000kg
Product Code. 11323922
  • 1970.00 SEK / Pack of 6
Estimated Shipment: 10-05-2024
to see stock.
Description
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip
Specifications
Specifications
26 G