Läs mer
Keto Bisoprolol Hydrochloride, TRC
Produktkod. 30408568
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30408568 10 mg
1 options
Produktkod. 30408568 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK17515510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Keto Bisoprolol Hydrochloride
CAS 1346603-26-6
Molekylformel C18H30ClNO5
Kvantitet 10 mg
Synonym 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-Benzoic Acid 2-(1-methylethoxy)ethyl Ester Hydrochloride, 2-Isopropoxyethyl-4-[[(2RS)-2-hydroxy-3-(isopropylamino)propyl]oxy]benzoate Hydrochloride, Bisoprolol Ester Hydrochloride Impurity
InChI formel InChI=1S/C18H29NO5.ClH/c1-13(2)19-11-16(20)12-24-17-7-5-15(6-8-17)18(21)23-10-9-22-14(3)4;/h5-8,13-14,16,19-20H,9-12H2,1-4H3;1H
LEDER CC(C)NCC(O)COc1ccc(cc1)C(=O)OCCOC(C)C
Molekylvikt (g/mol) 375.89