Läs mer
Ketotifen EP Impurity B, Mikromol™
Produktkod. 30052053
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052053 25 mg
30051052 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052053 Leverantör Mikromol Leverantörsnummer MM0180.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ketotifen EP Impurity B
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
CAS 59743-88-3
Analyt- eller komponentnamn (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
Frakt skick Room Temperature
Molekylformel C20 H23 N O2 S
Certifikat och efterlevnad ISO 17025
Synonym Ketotifen Hydrogen Fumarate Imp. B (EP), Ketotifen Imp. B (EP), (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol, Ketotifen Hydrogen Fumarate Impurity B, Ketotifen Impurity B
InChI formel InChI=1S/C20H23NO2S/c1-21-10-7-15(8-11-21)20(22)16-6-4-3-5-14(16)13-18(23-2)19-17(20)9-12-24-19/h3-6,9,12-13,15,22H,7-8,10-11H2,1-2H3
LEDER COC1=Cc2ccccc2C(O)(C3CCN(C)CC3)c4ccsc14
IUPAC-namn 4-methoxy-10-(1-methylpiperidin-4-yl)benzo[1,2]cyclohepta[2,4-b]thiophen-10-ol
Molekylvikt (g/mol) 341.47
Kvantitet 25 mg
Formel vikt 341.1449 g/mol