missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23443-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
KHDC1 family members are K-homology (KH) domain-containing RNA binding proteins that are involved in various aspects of RNA metabolism, ranging from transcription to RNA splicing, transportation, translation, and stability. KHDC1 is highly expressed in oocytes and interacts with cytoplasmic polyadenylation element-binding protein 1 (CPEB1), a key translational regulator controlling the polyA length of mRNAs. KHDC1 functions as a global translational repressor and induces apoptosis through an ER-dependent signaling pathway and that the C-terminal putative trans-membrane motif (TMM) is critical for its activities.Specifications
KHDC1 | |
Polyclonal | |
Unconjugated | |
KHDC1 | |
C6orf148, KHDC1, NDG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80759 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4VXA5 | |
KHDC1 | |
KHDC1 Fusion Protein Ag20115 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |