Läs mer
KHS101 hydrochloride, MedChemExpress
Produktkod. 16473036
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16473036 25 mg
16464338 10 mg
16437057 5 mg
16435256 100 mg
16420667 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16473036 Leverantör Medchem Express Leverantörsnummer HY10996A
MedChemExpress KHS101 hydrochloride could selectively induce a neuronal differentiation phenotype and interacts with transforming acidic coiled-coil-containing protein 3 (TACC3).
KHS101 hydrochloride could selectively induce a neuronal differentiation phenotype and interacts with transforming acidic coiled-coil-containing protein 3 (TACC3).
Specifikationer
Kemiskt namn eller material KHS101 hydrochloride
Fysisk form Solid
CAS 1784282-12-7
Färg White
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H22ClN5S
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 160 mg/mL (425.62 mM; Need ultrasonic) ∣H2O : 10 mg/mL (26.60 mM; ultrasonic and warming and heat to 60°C)
LEDER [H]Cl.CC(CNC1=NC(NCC2=CSC(C3=CC=CC=C3)=N2)=NC=C1)C
Molekylvikt (g/mol) 375.92
Formel vikt 375.92
Procent renhet 99.87%