missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki-67 Monoclonal Antibody (20Raj1), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 47-5699-41
Additional Details : Weight : 0.01000kg
Description
Ki-67 Monoclonal antibody specifically detects Ki-67 in Canine, Human samples. It is validated for Flow Cytometry.Specifications
Ki-67 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P46013 | |
Mki67 | |
Affinity chromatography | |
RUO | |
100686578, 4288 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
20Raj1 | |
APC-eFluor 780 | |
Mki67 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
Mouse | |
25 Tests | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 κ |