missing translation for 'onlineSavingsMsg'
Learn More

KIAA0284 Mouse anti-Human, Clone: 1D4B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66749-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828915

  • 1236.20 SEK / 20µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CEP170B plays a role in microtubule organization.
Specifications

Specifications

KIAA0284
1D4B8
Unconjugated
CEP170B
Liquid
CEP170B
KIAA0284 Fusion Protein Ag26852
Protein A
RUO
Antibody
Monoclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y4F5
Cep170B, FAM68C, KIAA0284, Protein KIAA0284
Mouse
IgG2b
20 μL
-20°C
Primary
283638
Videos
SDS
Documents
Certificates
Special Offers

Special Offers