missing translation for 'onlineSavingsMsg'
Learn More

KIAA0825 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA558215

Additional Details : Weight : 0.02000kg

Product Code. 15786091

  • 3500.00 SEK / 100µL


Estimated Shipment date
from Supplier 05-09-2019
Add to basket

Specifications

Specifications

KIAA0825
Polyclonal
Unconjugated
KIAA0825
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
285600
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IV33
KIAA0825
Recombit protein corresponding to Human KIAA0825
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documents

Research Use Only