missing translation for 'onlineSavingsMsg'
Learn More

KIAA0841 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25721-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884275

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

KIAA0841
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O94927
HAUS5, KIAA0841
Rabbit
IgG
20 μL
-20°C
Primary
23354
Immunohistochemistry (Paraffin)
Unconjugated
HAUS5
Liquid
HAUS5
KIAA0841 Fusion Protein Ag22809
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers