missing translation for 'onlineSavingsMsg'
Learn More

KIAA1033 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51101-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16896385

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a component of the WASH complex, which functions in the intracellular transport of endosomes. Mutations in this gene have been detected in individuals with autosomal recessive cognitive disability. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

KIAA1033
Polyclonal
Unconjugated
WASHC4
KIAA1033, SWIP, WASH complex subunit 7
Rabbit
Antigen Affinity Chromatography
RUO
23325
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q2M389
WASHC4
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers