Läs mer
Kif15-IN-1, MedChemExpress
Produktkod. 16469467
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16469467 5 mg
16437006 100 mg
16416928 10 mg
16402987 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16469467 Leverantör Medchem Express Leverantörsnummer HY15948
MedChemExpress Kif15-IN-1 is an inhibitor of the mitotic Kinesin family member 15 (Kif15), and is used for the research of cellular proliferative diseases.
Kif15-IN-1 is an inhibitor of the mitotic Kinesin family member 15 (Kif15), and is used for the research of cellular proliferative diseases.
Specifikationer
Kemiskt namn eller material Kif15-IN-1
Fysisk form Solid
CAS 672926-32-8
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H22N4O5S
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : ≥ 100 mg/mL (232.30 mM)
LEDER O=C(C1=CC=CC=C1N2)N([C@@H](C(C)C)C(NC3=NC(C)=C(C(OCC)=O)S3)=O)C2=O
Molekylvikt (g/mol) 430.48
Formel vikt 430.48
Procent renhet 99.23%