Läs mer
Kif15-IN-2, MedChemExpress
Produktkod. 16479467
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16479467 5 mg -
16447006 100 mg -
16426928 10 mg -
16412987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16479467 Leverantör Medchem Express Leverantörsnummer HY159495mg
MedChemExpress Kif15-IN-2 is an inhibitor of the mitotic kinesin Kif15, and is used for the research of cellular proliferative diseases.
Kif15-IN-2 is an inhibitor of the mitotic kinesin Kif15, and is used for the research of cellular proliferative diseases.
Specifikationer
Kemiskt namn eller material Kif15-IN-2
Fysisk form Solid
CAS 672926-33-9
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H20N6O4S
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 20 mg/mL (45.41 mM; Need ultrasonic)
LEDER O=C(C1=CC=CC=C1N2)N([C@@H](C(C)C)C(NC3=NC(C)=C(C4=NN=C(C)O4)S3)=O)C2=O
Molekylvikt (g/mol) 440.48
Formel vikt 440.48
Procent renhet 98.18%