missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KIF1B Recombinant Protein
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH01415 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 23095 |
Molecular Weight (g/mol) | 89.21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136942
|
Abnova™
H00023095-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16146942
|
Abnova™
H00023095-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01415 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
89.21 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIF1B | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
23095 | |
KIF1B (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSKTSLGQSMSKYDLLVWFEISELEPTGEYIPAVVDHTAGLPCQGTFLLHQGIQRRITVTIIHEKGSELHWKDVRELVVGRIRNKPEVDEAAVDAILSLNIISAKYLKSSHNSSRTFYRFEAVWDSSLHNSLLLNRVTPYGEKIYMTLSAYLELDHCIQPAVITKDVCMVFYSRDAKISPPRSLRSLFGSGYSKSPDSNRVTGIYELSLCKMSDTGSPGMQRRRRKILDTSVAYVRGEENLAGWRPRGDSLILEHQWELEKLELLHEVEKTRHFLLLRERLGDSIPKSLSDSLSPSLSSGTLSTSTSISSQISTTTFESAITPSESSGYDSGDIESLVDREKELATKCLQLLTHTFNREFSQVHGSVSDCKLSDISPIGRDPSESSFSSATLTPSSTCPSLVDSRSNSLDQKTPEANSRASSPCPEFEQFQIVPAVETPYLARAGKNEFLNLVPDIEEIRPSSVVSKKGYLHFKEPLYSNWAKHFVVVRRPYVFIYNSDKDPVERGIINLSTAQVEYSEDQQAMVKTPNTFAVCTKHRGVLLQALNDKDMNDWLYAFNPLLAGTIRSKLSRRCPSQSKY | |
CMT2/CMT2A/CMT2A1/FLJ23699/HMSNII/KIAA0591/KIAA1448/KLP/MGC134844 | |
KIF1B | |
Wheat Germ (in vitro) | |
GST |