missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF26B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17422-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
KIF26B is a member of kinesin-11 family and plays important role in embryogenesis. It is a downstream target of Sall1 and is essential for kidney development. KIF26B is also involved in tumorigenesis.Specifications
KIF26B | |
Polyclonal | |
Unconjugated | |
KIF26B | |
KIF26B, kinesin family member 26B, Kinesin like protein KIF26B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269152, 305012, 55083 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2KJY2, Q7TNC6 | |
Kif26b | |
KIF26B Fusion Protein Ag11446 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |