Learn More
KIF2C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28372-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The KIF2C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF2C. This antibody recognizes human, mouse , rat antigen. The KIF2C antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
KIF2C | |
Polyclonal | |
Unconjugated | |
150 μL | |
11004 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |