Learn More
KIF2C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28372-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16876115
  • 4721.67 SEK / 150µL
Estimated Shipment: 20-05-2024
to see stock.
Description
Description
The KIF2C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF2C. This antibody recognizes human, mouse , rat antigen. The KIF2C antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
KIF2C
Polyclonal
Unconjugated
150 μL
11004
IgG
Western Blot, Immunohistochemistry, ELISA
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse