missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12700-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 ′Armadillo′ repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. Several transcript variants encoding different isoforms have been found for this gene.Specifications
KIFAP3 | |
Polyclonal | |
Unconjugated | |
KIFAP3 | |
KAP 3, KAP3, KIF3AP, KIFAP3, kinesin associated protein 3, SMAP, Smg GDS, Smg GDS associated protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16579, 22920, 289168 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70188, Q92845 | |
Kifap3 | |
KIFAP3 Fusion Protein Ag3394 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |