missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20790-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody was raised against the C-terminal region of human KIFC1 protein, and recognizes 55-70 kDa of endogenous KIFC1 protein in multiple tissue lysates.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.Specifications
KIFC1 | |
Polyclonal | |
Unconjugated | |
KIFC1 | |
HSET, KIFC1, kinesin family member C1, Kinesin like protein 2, Kinesin like protein KIFC1, Kinesin related protein HSET, KNSL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100502766, 294286, 3833 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XI63, Q9BW19, Q9QWT9 | |
KIFC1 | |
KIFC1 Fusion Protein Ag14492 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |