Learn More
DWK Life Sciences Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock
Product Code. 12201368
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
12201368 50 mL Each
1 options
Product Code. 12201368 Supplier DWK Life Sciences Supplier No. 17027F50
  • ONLINE EXCLUSIVE PRICE
    3555.00 SEK
    3264.79 SEK / Paket med 1
    Save 290.21 SEK
    8% Av
 Restricted Item
Beräknad leverans: 21-01-2027
Includes
No. 2 PTFE stopcock and KimKap™ dust cap.
Class A
  • Precision bore tubing
  • Durable white scale
  • Permanently marked with individual serial number
  • Sold in a Case of 1
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.
Compliance
  • Meet ASTM™ E 287
  • 4 1/2 in. (115mm) delivery stem on 10mL size meets ASTM D 664
Specifications
Capacity (Metric) 50 mL
Length (Metric) 745 mm
Material Borosilicate Glass
Graduations 0.1 mL
For Use With (Application) General Purpose Titrations Requiring Traceable Volumetric Accuracy
Length (English) 29.33 in.
Includes Dust Cap
Scale White
Stopcock Straight
Stopcock Material PTFE
Tolerance ±0.05 mL
Volume (Metric) Reservoir 0.1 mL
Type Bore Buret
Class Class A
Certifications/Compliance ASTM E287, ASTM E438, ASTM E542, ASTM E694